Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336459.1 | NP_001325202.1 | 10 - 11 | 4179 | 100 | |
NM_128914.3 | NP_180912.2 | 11 - 12 | 4054 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |