Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036094.1 | NP_001031171.1 | 6 - 7 | 902 | 127 | |
NM_104037.4 | NP_175570.1 | 6 - 7 | 906 | 127 | ||
GenBank mRNA | AY081353.1 | - | 6 - 7 | 893 | 127 | |
AY128845.1 | - | 6 - 7 | 720 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |