Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSP41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116179.5 | NP_191873.1 | 4 - 5 | 927 | 67 | |
GenBank mRNA | AF428269.1 | - | 4 - 5 | 786 | 67 | |
AY059887.1 | - | 4 - 5 | 849 | 67 | ||
AY128804.1 | - | 4 - 5 | 758 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |