Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036494.1 | NP_001031571.1 | 2 - 3 | 290 | 132 | |
NM_001125441.1 | NP_001118913.1 | 2 - 3 | 418 | 132 | ||
NM_116398.4 | NP_192077.1 | 2 - 3 | 442 | 132 | ||
GenBank mRNA | AF176000.2 | - | 2 - 3 | 254 | 132 | |
AY054575.1 | - | 2 - 3 | 410 | 132 | ||
BT000352.1 | - | 2 - 3 | 254 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |