Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036500.2 | NP_001031577.1 | 5 - 6 | 886 | 80 | |
NM_001125447.1 | NP_001118919.1 | 6 - 7 | 644 | 80 | ||
NM_116493.3 | NP_567245.1 | 5 - 6 | 935 | 80 | ||
GenBank mRNA | AF367328.1 | - | 5 - 6 | 816 | 80 | |
AK316748.1 | - | 5 - 6 | 887 | 80 | ||
AY143951.1 | - | 5 - 6 | 701 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |