Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hemf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116557.4 | NP_567256.3 | 1 - 2 | 556 | 58 | |
NM_116558.2 | NP_567257.2 | 1 - 2 | 556 | 58 | ||
GenBank mRNA | AY057708.1 | - | 1 - 2 | 556 | 58 | |
BT006346.1 | - | 1 - 2 | 527 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |