Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203749.1 | NP_001190678.1 | 10 - 11 | 1906 | 100 | |
NM_116741.4 | NP_567283.1 | 9 - 10 | 1989 | 100 | ||
GenBank mRNA | AY039856.1 | - | 9 - 10 | 1721 | 100 | |
BT002241.1 | - | 9 - 10 | 1664 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |