Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116971.6 | NP_192641.2 | 23 - 24 | 2371 | 55 | |
GenBank mRNA | AK227049.1 | - | 23 - 24 | 2208 | 55 | |
AY091058.1 | - | 23 - 24 | 2204 | 55 | ||
AY133739.1 | - | 23 - 24 | 2181 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |