Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116972.5 | NP_192642.1 | 1 | 407 | 76 | |
GenBank mRNA | AF195891.1 | - | 1 | 322 | 76 | |
AK229123.1 | - | 1 | 337 | 76 |
Gene Symbol: AT4G09035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340609.1 | NP_001329944.1 | 1 | 83 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |