Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333655.1 | NP_001320715.1 | 34 - 35 | 4581 | 125 | |
NM_104334.2 | NP_175858.1 | 33 - 34 | 4305 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |