Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104613.4 | NP_564730.1 | 1 | 893 | 85 | |
GenBank mRNA | AB028196.1 | - | 1 | 854 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |