Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCF61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333855.1 | NP_001320817.1 | 4 - 5 | 546 | 92 | |
NM_001333856.1 | NP_001320816.1 | 4 - 5 | 504 | 92 | ||
NM_001333857.1 | NP_001320818.1 | 4 - 5 | 481 | 92 | ||
NM_104650.4 | NP_564745.1 | 4 - 5 | 546 | 92 | ||
GenBank mRNA | AK228076.1 | - | 4 - 5 | 481 | 92 | |
BT006461.1 | - | 4 - 5 | 339 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |