Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104914.2 | NP_564793.2 | 7 - 8 | 1565 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |