Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKU5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203775.2 | NP_001190704.1 | 6 - 7 | 1656 | 64 | |
NM_117312.4 | NP_192979.1 | 6 - 7 | 1638 | 64 | ||
GenBank mRNA | AF439406.1 | - | 6 - 7 | 1389 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |