Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129043.5 | NP_181036.2 | 6 - 7 | 865 | 66 | |
NM_201873.2 | NP_973602.1 | 6 - 7 | 812 | 66 | ||
GenBank mRNA | AK230434.1 | - | 6 - 7 | 827 | 66 | |
AY180100.1 | - | 6 - 7 | 743 | 66 | ||
BT026469.1 | - | 6 - 7 | 743 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |