Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110984.3 | NP_186767.1 | 6 - 7 | 1680 | 67 | |
GenBank mRNA | AY054467.1 | - | 6 - 7 | 1348 | 67 | |
BT002555.1 | - | 6 - 7 | 1247 | 67 | ||
KJ138848.1 | - | 6 - 7 | 1247 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |