Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340852.1 | NP_001328594.1 | 5 - 6 | 1008 | 147 | |
NM_001340853.1 | NP_001328593.1 | 5 - 6 | 977 | 147 | ||
NM_001340854.1 | NP_001328592.1 | 5 - 6 | 947 | 147 | ||
NM_001340855.1 | NP_001328591.1 | 5 - 6 | 1436 | 147 | ||
NM_001340856.1 | NP_001328590.1 | 5 - 6 | 1311 | 147 | ||
NM_117408.4 | NP_567403.1 | 5 - 6 | 1008 | 147 | ||
NM_179042.1 | NP_849373.1 | 5 - 6 | 947 | 147 | ||
GenBank mRNA | AY059783.1 | - | 5 - 6 | 946 | 147 | |
AY117270.1 | - | 4 - 5 | 447 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |