Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117409.5 | NP_193071.2 | 4 - 5 | 530 | 95 | |
NM_179209.2 | NP_849540.1 | 4 - 5 | 378 | 95 | ||
GenBank mRNA | AY062505.1 | - | 4 - 5 | 378 | 95 | |
BT008379.1 | - | 3 - 4 | 248 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |