Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCOAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105468.3 | NP_564916.2 | 2 - 3 | 181 | 69 | |
NM_202375.1 | NP_974104.1 | 2 - 3 | 149 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |