Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036196.2 | NP_001031273.1 | 16 - 17 | 2109 | 154 | |
NM_001334559.1 | NP_001321344.1 | 16 - 17 | 2104 | 154 | ||
NM_001334560.1 | NP_001321343.1 | 17 - 18 | 1961 | 154 | ||
NM_105957.3 | NP_565051.1 | 16 - 17 | 2096 | 154 | ||
GenBank mRNA | AY058198.1 | - | 16 - 17 | 1998 | 154 | |
AY142008.1 | - | 16 - 17 | 1777 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |