Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334569.1 | NP_001320310.1 | 2 - 3 | 1625 | 117 | |
NM_105973.4 | NP_177458.1 | 3 - 4 | 1747 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |