Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344127.1 | NP_001318680.1 | 12 - 13 | 1695 | 107 | |
NM_123006.4 | NP_198464.4 | 11 - 12 | 1629 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |