Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344166.1 | NP_001318683.1 | 10 - 11 | 2342 | 73 | |
NM_001344167.1 | NP_001332704.1 | 6 - 7 | 1747 | 73 | ||
NM_123077.3 | NP_198534.3 | 8 - 9 | 1922 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |