Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339129.1 | NP_001326943.1 | 24 - 25 | 3517 | 113 | |
NM_001339130.1 | NP_001326945.1 | 25 - 26 | 3418 | 113 | ||
NM_114302.3 | NP_566869.1 | 25 - 26 | 3388 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |