Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339152.1 | NP_001319682.1 | 7 - 8 | 1790 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |