Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111345.3 | NP_187124.1 | 4 - 5 | 1151 | 122 | |
GenBank mRNA | AF375420.1 | - | 4 - 5 | 858 | 122 | |
AK229447.1 | - | 4 - 5 | 858 | 122 | ||
AY087195.1 | - | 4 - 5 | 862 | 122 | ||
U49072.1 | - | 4 - 5 | 841 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |