Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIIIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341848.1 | NP_001328308.1 | 11 - 12 | 1425 | 100 | |
NM_001341849.1 | NP_001328309.1 | 10 - 11 | 1364 | 100 | ||
NM_001341850.1 | NP_001328307.1 | 10 - 11 | 1441 | 100 | ||
NM_118871.6 | NP_194467.5 | 9 - 10 | 1232 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |