Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119096.3 | NP_194680.1 | 6 - 7 | 823 | 113 | |
GenBank mRNA | AY087817.1 | - | 6 - 7 | 820 | 113 | |
AY150407.1 | - | 6 - 7 | 706 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |