Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345090.1 | NP_001330204.1 | 2 - 3 | 213 | 71 | |
NM_124837.3 | NP_200268.3 | 2 - 3 | 213 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |