Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124872.3 | NP_568815.1 | 5 - 6 | 1054 | 77 | |
GenBank mRNA | AF324983.2 | - | 5 - 6 | 1040 | 77 | |
BT029545.1 | - | 5 - 6 | 1011 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |