Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100191.4 | NP_563674.1 | 10 - 11 | 1561 | 85 | |
NM_179252.3 | NP_849583.1 | 11 - 12 | 1755 | 85 | ||
GenBank mRNA | AY070723.1 | - | 11 - 12 | 1612 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |