Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035924.2 | NP_001031001.2 | 4 - 5 | 686 | 65 | |
NM_001084023.1 | NP_001077492.1 | 4 - 5 | 726 | 65 | ||
NM_001331780.1 | NP_001318956.1 | 3 - 4 | 697 | 65 | ||
NM_001331781.1 | NP_001322023.1 | 4 - 5 | 647 | 65 | ||
NM_100736.3 | NP_563822.1 | 4 - 5 | 1163 | 65 | ||
NM_179283.2 | NP_849614.1 | 4 - 5 | 726 | 65 | ||
NM_179284.4 | NP_849615.1 | 4 - 5 | 759 | 65 | ||
GenBank mRNA | AK230078.1 | - | 4 - 5 | 768 | 65 | |
AK317034.1 | - | 4 - 5 | 691 | 65 | ||
AY080670.1 | - | 4 - 5 | 722 | 65 | ||
AY117227.1 | - | 2 - 3 | 557 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |