Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123785.1 | NP_001117257.1 | 2 - 3 | 982 | 69 | |
NM_001331880.1 | NP_001320930.1 | 4 - 5 | 856 | 69 | ||
NM_001331881.1 | NP_001320929.1 | 4 - 5 | 856 | 69 | ||
NM_001331882.1 | NP_001320928.1 | 4 - 5 | 819 | 69 | ||
NM_001331883.1 | NP_001320927.1 | 4 - 5 | 690 | 69 | ||
NM_100880.3 | NP_563859.1 | 4 - 5 | 690 | 69 | ||
NM_179298.2 | NP_849629.1 | 4 - 5 | 690 | 69 | ||
GenBank mRNA | AK316768.1 | - | 4 - 5 | 619 | 69 | |
AY054517.1 | - | 4 - 5 | 691 | 69 | ||
AY128778.1 | - | 4 - 5 | 484 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |