Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202945.2 | NP_001189874.1 | 5 - 6 | 725 | 87 | |
NM_001338017.1 | NP_001319535.1 | 5 - 6 | 649 | 87 | ||
NM_001338018.1 | NP_001327698.1 | 5 - 6 | 649 | 87 | ||
NM_112136.5 | NP_566441.1 | 4 - 5 | 705 | 87 | ||
GenBank mRNA | AK227231.1 | - | 4 - 5 | 705 | 87 | |
BT003077.1 | - | 4 - 5 | 558 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |