Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSPO11-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338034.1 | NP_001326966.1 | 10 - 11 | 651 | 103 | |
NM_001338035.1 | NP_001326967.1 | 10 - 11 | 637 | 103 | ||
NM_112156.2 | NP_187923.1 | 10 - 11 | 651 | 103 | ||
GenBank mRNA | AJ251989.1 | - | 10 - 11 | 690 | 103 | |
BT028955.1 | - | 10 - 11 | 637 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |