Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKKK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112199.3 | NP_187962.1 | 22 - 23 | 7497 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |