Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119316.5 | NP_194895.1 | 7 - 8 | 1162 | 124 | |
GenBank mRNA | AY142532.1 | - | 7 - 8 | 1055 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |