Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: La1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036700.2 | NP_001031777.1 | 5 - 6 | 677 | 99 | |
NM_119425.5 | NP_567904.1 | 5 - 6 | 692 | 99 | ||
GenBank mRNA | AY056237.1 | - | 5 - 6 | 616 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |