Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT4G34840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342303.1 | NP_001329682.1 | 5 - 6 | 439 | 121 | |
NM_001342304.1 | NP_001329684.1 | 5 - 6 | 472 | 121 | ||
NM_001342305.1 | NP_001329685.1 | 4 - 5 | 539 | 121 | ||
NM_001342306.1 | NP_001329683.1 | 5 - 6 | 472 | 121 | ||
NM_119650.4 | NP_195210.2 | 5 - 6 | 472 | 121 | ||
GenBank mRNA | AK229730.1 | - | 5 - 6 | 445 | 121 | |
BT010195.1 | - | 5 - 6 | 395 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |