Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334865.1 | NP_001321029.1 | 7 - 8 | 1055 | 151 | |
NM_106546.3 | NP_178018.1 | 7 - 8 | 1159 | 151 | ||
NM_179572.2 | NP_849903.1 | 6 - 7 | 997 | 151 | ||
GenBank mRNA | AF360147.1 | - | 6 - 7 | 987 | 151 | |
AY052358.1 | - | 7 - 8 | 1150 | 151 | ||
AY142678.1 | - | 6 - 7 | 913 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |