Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036292.2 | NP_001031369.1 | 2 - 3 | 810 | 103 | |
NM_179644.2 | NP_849975.1 | 2 - 3 | 777 | 103 | ||
GenBank mRNA | AK317092.1 | - | 2 - 3 | 734 | 103 | |
AY045773.1 | - | 1 - 2 | 332 | 103 | ||
BT025314.1 | - | 1 - 2 | 332 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |