Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335657.1 | NP_001323708.1 | 4 - 5 | 731 | 94 | |
NM_179663.3 | NP_849994.1 | 4 - 5 | 731 | 94 | ||
GenBank mRNA | AJ487669.1 | - | 4 - 5 | 731 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |