Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL1-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112481.4 | NP_188232.2 | 7 - 8 | 1400 | 68 | |
GenBank mRNA | BT015349.1 | - | 7 - 8 | 1222 | 68 | |
BT015798.1 | - | 7 - 8 | 1167 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |