Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084702.2 | NP_001078171.1 | 3 - 4 | 520 | 77 | |
GenBank mRNA | AK176588.1 | - | 3 - 4 | 315 | 77 | |
AK176702.1 | - | 3 - 4 | 325 | 77 | ||
AK176807.1 | - | 3 - 4 | 325 | 77 | ||
BT015109.1 | - | 3 - 4 | 288 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |