Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112690.4 | NP_188436.1 | 6 - 7 | 921 | 85 | |
GenBank mRNA | AK316840.1 | - | 6 - 7 | 824 | 85 | |
AK316900.1 | - | 6 - 7 | 825 | 85 | ||
AY084864.1 | - | 6 - 7 | 823 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |