Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGSIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035645.3 | NP_001030722.1 | 3 - 4 | 1120 | 77 | |
NM_001202998.1 | NP_001189927.1 | 3 - 4 | 966 | 77 | ||
NM_112752.3 | NP_566615.1 | 3 - 4 | 950 | 77 | ||
GenBank mRNA | AY056133.1 | - | 3 - 4 | 950 | 77 | |
AY142690.1 | - | 3 - 4 | 926 | 77 | ||
AY749109.1 | - | 3 - 4 | 938 | 77 | ||
KJ138819.1 | - | 3 - 4 | 938 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |