Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112829.5 | NP_188573.1 | 4 - 5 | 952 | 115 | |
GenBank mRNA | AY070042.1 | - | 4 - 5 | 817 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |