Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035654.2 | NP_001030731.1 | 1 - 2 | 216 | 106 | |
NM_001338474.1 | NP_001327849.1 | 2 - 3 | 294 | 106 | ||
NM_112920.4 | NP_566657.1 | 2 - 3 | 361 | 106 | ||
GenBank mRNA | AK317308.1 | - | 1 - 2 | 282 | 106 | |
AY034911.1 | - | 2 - 3 | 218 | 106 | ||
BT004414.1 | - | 1 - 2 | 88 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |