Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLGAMMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203013.1 | NP_001189942.1 | 3 - 4 | 898 | 69 | |
NM_112946.5 | NP_188690.3 | 2 - 3 | 1032 | 69 | ||
GenBank mRNA | AB211533.1 | - | 2 - 3 | 825 | 69 | |
AK226811.1 | - | 2 - 3 | 958 | 69 | ||
AY195962.1 | - | 2 - 3 | 901 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |