Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119941.4 | NP_195493.1 | 1 - 2 | 648 | 127 | |
GenBank mRNA | AJ441254.1 | - | 1 - 2 | 449 | 127 | |
AY087563.1 | - | 1 - 2 | 554 | 127 | ||
BT002318.1 | - | 1 - 2 | 449 | 127 | ||
U09336.1 | - | 1 - 2 | 530 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |